Anti-MYOC Mouse Monoclonal Antibody [clone: 2B4]
Lieferant: Abnova
ABNOH00004653-M02EA 394 EUR
ABNOH00004653-M02
Anti-MYOC Mouse Monoclonal Antibody [clone: 2B4]
Antikörper
Type: Primary
Antigen: MYOC
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human