Anti-MYOD Rabbit Polyclonal Antibody
Lieferant: Abnova
ABNOH00004654-D01PEA 380 EUR
ABNOH00004654-D01P
Anti-MYOD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MYOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse