Anti-NAB2 Mouse Monoclonal Antibody [clone: 4H6]

Lieferant: Abnova
ABNOH00004665-M01EA 394 EUR
ABNOH00004665-M01
Anti-NAB2 Mouse Monoclonal Antibody [clone: 4H6]
Antikörper
Type: Primary
Antigen: NAB2
Clonality: Monoclonal
Clone: 4H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR