Anti-NAGLU Mouse Monoclonal Antibody [clone: 1B7]

Lieferant: Abnova
ABNOH00004669-M02EA 394 EUR
ABNOH00004669-M02
Anti-NAGLU Mouse Monoclonal Antibody [clone: 1B7]
Antikörper
Type: Primary
Antigen: NAGLU
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR