Anti-HNRNPM Mouse Monoclonal Antibody [clone: 2B6]
Lieferant: Abnova
ABNOH00004670-M01EA 394 EUR
ABNOH00004670-M01
Anti-HNRNPM Mouse Monoclonal Antibody [clone: 2B6]
Antikörper
Type: Primary
Antigen: HNRNPM
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human