Anti-NAPL1 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00004673-B02PEA 394 EUR
ABNOH00004673-B02P
Anti-NAPL1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR