Anti-RPL19 Mouse Monoclonal Antibody [clone: 3H4]
Lieferant: Abnova
ABNOH00006143-M01EA 394 EUR
ABNOH00006143-M01
Anti-RPL19 Mouse Monoclonal Antibody [clone: 3H4]
Antikörper
Type: Primary
Antigen: RPL19
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat