Anti-GSTCD Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB3008-100EA 312 EUR
BIRBORB3008-100 BIRBORB3008-200
Anti-GSTCD Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: GSTCD (glutathione S-transferase, C-terminal domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR