Anti-GTDC1 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB3010-100EA 0 EUR
BIRBORB3010-100 BIRBORB3010-200
Anti-GTDC1 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: GTDC1 (glycosyltransferase-like domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human