Anti-C4orf19 Rabbit polyclonal antibody

Lieferant: Biorbyt
BIRBORB3012-200EA 508 EUR
BIRBORB3012-200 BIRBORB3012-100
Anti-C4orf19 Rabbit polyclonal antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: C4orf19 (chromosome 4 open reading frame 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR