Anti-NINJ2 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB3022-100EA 0 EUR
BIRBORB3022-100 BIRBORB3022-200
Anti-NINJ2 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: NINJ2 (ninjurin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat