Anti-PCDH11X Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB3023-200EA 0 EUR
BIRBORB3023-200 BIRBORB3023-100
Anti-PCDH11X Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: PCDH11X (protocadherin 11 X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR