Anti-PCDH1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB3027-100EA 312 EUR
BIRBORB3027-100 BIRBORB3027-200
Anti-PCDH1 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: PCDH1 (protocadherin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR