Anti-PAFAH1B2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Biorbyt
BIRBORB3139-100EA 0 EUR
BIRBORB3139-100
Anti-PAFAH1B2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: PAFAH1B2 (platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat