Anti-KLF1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Biorbyt
BIRBORB3331-100EA 385 EUR
BIRBORB3331-100
Anti-KLF1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: KLF1 (Kruppel-like factor 1 (erythroid))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR