Anti-RNF32 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Biorbyt
BIRBORB3503-100EA 385 EUR
BIRBORB3503-100
Anti-RNF32 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: RNF32 (ring finger protein 32)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR