Anti-ANKRD45 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Biorbyt
BIRBORB3629-100EA 0 EUR
BIRBORB3629-100
Anti-ANKRD45 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: ANKRD45 (ankyrin repeat domain 45)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat