Anti-Myosin IIA Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Biorbyt
BIRBORB4013-100EA 0 EUR
BIRBORB4013-100
Anti-Myosin IIA Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: Myosin IIA
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen