Anti-IgG1 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Biorbyt
BIRBORB21741-1EA 0 EUR
BIRBORB21741-1
Anti-IgG1 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: Rat IgG1
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR