Anti-IgA Goat Polyclonal Antibody (Biotin)

Lieferant: Biorbyt
BIRBORB21824-1EA 0 EUR
BIRBORB21824-1
Anti-IgA Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: Mouse IgA
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR