Anti-kappa light chain Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB21908-10EA 0 EUR
BIRBORB21908-10
Anti-kappa light chain Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR