Anti-LysozymeLysozyme Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB21930-10EA 0 EUR
BIRBORB21930-10
Anti-LysozymeLysozyme Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: Lysozyme
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR