Anti-Secretory Component Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Biorbyt
BIRBORB21951-1EA 0 EUR
BIRBORB21951-1
Anti-Secretory Component Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: Secretory Component
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR