Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Biorbyt
BIRBORB21976-1EA 0 EUR
BIRBORB21976-1
Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: J Chain Of Dimeric IgA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR