Anti-Lactoferrin Goat Polyclonal Antibody (Biotin)

Lieferant: Biorbyt
BIRBORB22008-1EA 0 EUR
BIRBORB22008-1
Anti-Lactoferrin Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: Lactoferrin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR