Anti-IgD Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22158-1EA 0 EUR
BIRBORB22158-1
Anti-IgD Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: Mouse IgD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR