Anti-IgA1 Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB22217-0.2EA 0 EUR
BIRBORB22217-0.2
Anti-IgA1 Mouse Monoclonal Antibody
Antikörper
Type: Secondary
Antigen: IgA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR