Anti-IgG3 Mouse Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Biorbyt
BIRBORB22272-0.2EA 0 EUR
BIRBORB22272-0.2
Anti-IgG3 Mouse Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG3
Clonality: Monoclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR