Anti-IgA Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22297-1EA 0 EUR
BIRBORB22297-1
Anti-IgA Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgA (Cat) (cat IgA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR