Anti-IgM Pig Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22463-1EA 0 EUR
BIRBORB22463-1
Anti-IgM Pig Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Pig
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR