Anti-IRF2BP2 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22524-100EA 0 EUR
BIRBORB22524-100
Anti-IRF2BP2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IRF2BP2 (interferon regulatory factor 2 binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR