Anti-ASNSD1 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22548-100EA 0 EUR
BIRBORB22548-100
Anti-ASNSD1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ASNSD1 (asparagine synthetase domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR