Anti-BHMT Mouse monoclonal antibody

Lieferant: Biorbyt
BIRBORB22755-100EA 0 EUR
BIRBORB22755-100 BIRBORB22755-5 BIRBORB22755-20
Anti-BHMT Mouse monoclonal antibody
Antikörper
Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR