Anti-NAT6 Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB22777-5EA 0 EUR
BIRBORB22777-5 BIRBORB22777-100 BIRBORB22777-20
Anti-NAT6 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NAT6 (N-acetyltransferase 6 (GCN5-related))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR