Anti-VIM Mouse Monoclonal Antibody (Biotin)

Lieferant: Biorbyt
BIRBORB22923-1EA 0 EUR
BIRBORB22923-1 BIRBORB22923-500
Anti-VIM Mouse Monoclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR