Anti-Toxoplasma gondii Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB23596-1EA 0 EUR
BIRBORB23596-1
Anti-Toxoplasma gondii Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Toxoplasma gondii
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR