Anti-Epstein Barr Virus (LMP) Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB24117-1EA 0 EUR
BIRBORB24117-1
Anti-Epstein Barr Virus (LMP) Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 1:100-1:200

Type: Primary
Antigen: Epstein Barr Virus (LMP)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR