Anti-TNFRSF11B Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB24164-100EA 0 EUR
BIRBORB24164-100
Anti-TNFRSF11B Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:200-1:1000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: TNFRSF11B (tumor necrosis factor receptor superfamily, member 11b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR