Anti-GHRL Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB24379-1EA 0 EUR
BIRBORB24379-1
Anti-GHRL Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR