Anti-INS Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB24429-1EA 0 EUR
BIRBORB24429-1
Anti-INS Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR