Anti-Vancomycin Sheep Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB24448-0.5EA 0 EUR
BIRBORB24448-0.5
Anti-Vancomycin Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Vancomycin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR