Anti-FABP Mouse monoclonal antibody

Lieferant: Biorbyt
BIRBORB24693-1EA 0 EUR
BIRBORB24693-1
Anti-FABP Mouse monoclonal antibody
Antikörper
Type: Primary
Antigen: FABP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR