Anti-Trypsin Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB24773-0.2EA 0 EUR
BIRBORB24773-0.2
Anti-Trypsin Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Trypsin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR