Anti-glyceraldehyde-3-PDH Mouse monoclonal antibody

Lieferant: Biorbyt
BIRBORB24832-1EA 0 EUR
BIRBORB24832-1
Anti-glyceraldehyde-3-PDH Mouse monoclonal antibody
Antikörper
Type: Primary
Antigen: glyceraldehyde-3-PDH
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR