Anti-TNNT2 Mouse Monoclonal Antibody [clone: 1A11]

Lieferant: Biorbyt
BIRBORB24994-1EA 0 EUR
BIRBORB24994-1
Anti-TNNT2 Mouse Monoclonal Antibody [clone: 1A11]
Antikörper
Type: Primary
Antigen: TNNT2 (troponin T type 2 (cardiac))
Clonality: monoclonal
Clone: 1A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR