Anti-MPO Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB25001-0.2EA 0 EUR
BIRBORB25001-0.2
Anti-MPO Mouse Monoclonal Antibody
Antikörper
Flow Cytometry: 1:50-1:100; Immunochemistry: 1:1000-1:5000; ELISA: 1:500-1:1000

Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR