Anti-THPO Sheep Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB25033-1EA 0 EUR
BIRBORB25033-1
Anti-THPO Sheep Polyclonal Antibody
Antikörper
ELISA: 1:50000; Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: THPO (thrombopoietin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR