Anti-APOA1 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB25281-1EA 0 EUR
BIRBORB25281-1
Anti-APOA1 Goat Polyclonal Antibody
Antikörper
ELISA: 1:10000-1:80000

Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR