Anti-APOA1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Biorbyt
BIRBORB25282-1EA 0 EUR
BIRBORB25282-1
Anti-APOA1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
ELISA: 1:200-1:8000

Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR