Anti-Trypsin Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB25305-1EA 0 EUR
BIRBORB25305-1
Anti-Trypsin Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:512; Western Blot: 1:2000

Type: Primary
Antigen: Trypsin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR